Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005164.3 | NP_005155.1 | 10 | 2579 | |
XM_011538027.2 | XP_011536329.1 | 11 | 2519 | ||
GenBank mRNA | AJ000327.1 | - | 10 | 2288 | |
AK314254.1 | - | 10 | 2579 | ||
BC104901.1 | - | 10 | 2185 | 8143904 | |
BC104903.1 | - | 10 | 2179 | 8143906 |