Gene Transcripts
- Gene Symbol
- CCL16
- Entrez Gene ID
- 6360
- Gene Name
- C-C motif chemokine ligand 16
- Gene Aliases
- CKb12, HCC-4, ILINCK, LCC-1, LEC, LMC, Mtn-1, NCC-4, NCC4, SCYA16, SCYL4
- Chromosome Location
- Chr. 17: 35976493 - 35983620 on Build GRCh38
- UniGene ID
- Hs.10458
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004590.3 |
NP_004581.1
|
2, 3 |
275 |
|
XM_005258020.4 |
XP_005258077.1
|
4, 5 |
557 |
|
GenBank mRNA |
AB007454.1 |
- |
2, 3 |
275 |
|
AF039955.1 |
- |
2, 3 |
266 |
|
AF055467.1 |
- |
2, 3 |
233 |
|
BC096352.4 |
- |
3, 4 |
471 |
|
BC099662.3 |
- |
2, 3 |
240 |
40009719 |
BC099663.1 |
- |
3, 4 |
471 |
|
BC099664.4 |
- |
3, 4 |
471 |
|
BQ722834.1 |
- |
1, 2 |
152 |
|
U91746.1 |
- |
2, 3 |
199 |
|
View Full Product Details