Gene Transcripts

Gene Symbol
CCL16
Entrez Gene ID
6360
Gene Name
C-C motif chemokine ligand 16
Gene Aliases
CKb12, HCC-4, ILINCK, LCC-1, LEC, LMC, Mtn-1, NCC-4, NCC4, SCYA16, SCYL4
Chromosome Location
Chr. 17: 35976493 - 35983620 on Build GRCh38
UniGene ID
Hs.10458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004590.3 NP_004581.1 3 386
XM_005258020.4 XP_005258077.1 5 668
GenBank mRNA AB007454.1 - 3 386
AF039955.1 - 3 377
AF055467.1 - 3 344
BC096352.4 - 4 582
BC099662.3 - 3 351 40009719
BC099663.1 - 4 582
BC099664.4 - 4 582
BQ722834.1 - 2 263
U91746.1 - 3 310

View Full Product Details