Gene Transcripts

Gene Symbol
Efr3b
Entrez Gene ID
668212
Gene Name
EFR3 homolog B (S. cerevisiae)
Gene Aliases
AI852640, C030014M07Rik, mKIAA0953
Chromosome Location
Chr. 12: 3962554 - 4039094 on Build GRCm38
UniGene ID
Mm.210266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082483.1 NP_001075952.1 6 806
GenBank mRNA AK129247.1 - 2 253
BC075650.1 - 5 563
BC147643.1 - 6 806 9056633
BC157968.1 - 6 806 9088136

View Full Product Details