Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007808.4 | NP_031834.1 | 2 | 152 | |
GenBank mRNA | AK002732.1 | - | 2 | 154 | |
AK003168.1 | - | 2 | 181 | ||
AK005336.1 | - | 2 | 224 | ||
AK010651.1 | - | 2 | 156 | ||
AK088098.1 | - | 2 | 153 | ||
AK143887.1 | - | 2 | 153 | ||
AK168017.1 | - | 2 | 152 | ||
BC034363.1 | - | 2 | 136 | 2650742 | |
BC092213.1 | - | 1 | 101 | 6845432 | |
BC094054.1 | - | 2 | 141 | 2865963 |