Gene Transcripts

Gene Symbol
SELPLG
Entrez Gene ID
6404
Gene Name
selectin P ligand
Gene Aliases
CD162, CLA, PSGL-1, PSGL1
Chromosome Location
Chr. 12: 108621895 - 108633894 on Build GRCh38
UniGene ID
Hs.591014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206609.1 NP_001193538.1 2 1052
NM_003006.4 NP_002997.2 2 987
GenBank mRNA AK098315.1 - 4 906
AK127638.1 - 3 938
AK225644.1 - 2 883
AK290395.1 - 3 924
AK295238.1 - 3 769
AK298738.1 - 2 985
AK298742.1 - 2 1024
AK300057.1 - 3 879
BC029782.1 - 3 936 5180824
U02297.1 - 3 856

View Full Product Details