Gene Transcripts

Gene Symbol
Fbp1
Entrez Gene ID
24362
Gene Name
fructose-1,6-bisphosphatase 1
Gene Aliases
Fdp
Chromosome Location
Chr. 17: 396175 - 417480 on Build Rnor_6.0
UniGene ID
Rn.33703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012558.3 NP_036690.2 7 1066
GenBank mRNA BC078894.1 - 7 1048
BC078895.1 - 7 1066 7134943
J04112.1 - 7 862
M86240.1 - 7 952

View Full Product Details