Gene Transcripts

Gene Symbol
Prkcb
Entrez Gene ID
25023
Gene Name
protein kinase C, beta
Gene Aliases
Pkcb, Prkcb1
Chromosome Location
Chr. 1: 192233780 - 192575339 on Build Rnor_6.0
UniGene ID
Rn.91118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012713.3 NP_036845.3 18 2085
GenBank mRNA CB576043.1 - 2 218
FQ233088.1 - 1 70
M13706.1 - 16 2095
X04440.1 - 16 2639

View Full Product Details