Gene Transcripts

Gene Symbol
SLC16A2
Entrez Gene ID
6567
Gene Name
solute carrier family 16 member 2
Gene Aliases
AHDS, DXS128, DXS128E, MCT 7, MCT 8, MCT7, MCT8, MRX22, XPCT
Chromosome Location
Chr. X: 74421493 - 74533929 on Build GRCh38
UniGene ID
Hs.75317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006517.4 NP_006508.2 1 491
GenBank mRNA AB085789.1 - 1 433
CN342861.1 - 1 493
U05315.1 - 1 523

View Full Product Details