Gene Transcripts

Gene Symbol
Uncx
Entrez Gene ID
29375
Gene Name
UNC homeobox
Gene Aliases
Chx4, PHD1, Uncx4.1
Chromosome Location
Chr. 12: 17182158 - 17186679 on Build Rnor_6.0
UniGene ID
Rn.10710
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017179.1 NP_058875.1 2, 3 965
GenBank mRNA D87748.1 - 2, 3 965

View Full Product Details