Gene Transcripts

Gene Symbol
Psmb5
Entrez Gene ID
29425
Gene Name
proteasome (prosome, macropain) subunit, beta type 5
Gene Aliases
Chromosome Location
Chr. 15: 33259039 - 33263634 on Build Rnor_6.0
UniGene ID
Rn.2
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105727.2 NP_001099197.2 2 297
GenBank mRNA CB783917.1 - 2 297
D45247.1 - 2 295

View Full Product Details