Gene Transcripts

Gene Symbol
Uchl1
Entrez Gene ID
29545
Gene Name
ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)
Gene Aliases
Chromosome Location
Chr. 14: 43133224 - 43143942 on Build Rnor_6.0
UniGene ID
Rn.107213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017237.3 NP_058933.2 4 261
GenBank mRNA BC060573.1 - 4 265
D10699.1 - 4 245
FQ214229.1 - 4 257

View Full Product Details