Gene Transcripts
- Gene Symbol
- Crabp2
- Entrez Gene ID
- 29563
- Gene Name
- cellular retinoic acid binding protein 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 184882970 - 184883457 on Build Rnor_6.0
- UniGene ID
- Rn.11333
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017244.1 |
NP_058940.1
|
|
397 |
|
GenBank mRNA |
U23407.1 |
- |
|
397 |
|
- Gene Symbol
- LOC100911902
- Entrez Gene ID
- 100911902
- Gene Name
- cellular retinoic acid-binding protein 2-like
- Gene Aliases
- Chromosome Location
- Chr. 2: 187321661 - 187326794 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details