Gene Transcripts

Gene Symbol
SNRPC
Entrez Gene ID
6631
Gene Name
small nuclear ribonucleoprotein polypeptide C
Gene Aliases
U1C, Yhc1
Chromosome Location
Chr. 6: 34757094 - 34773857 on Build GRCh38
UniGene ID
Hs.1063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003093.2 NP_003084.1 3 560
NR_029472.1 2 517
GenBank mRNA AK311955.1 - 3 140
BC121082.1 - 3 129 40119776
BC121083.1 - 3 129 40119777
BU153661.1 - 2 125
BU508721.1 - 2 86
CD109745.1 - 3 577
M18465.1 - 1 38
X12517.1 - 3 125

View Full Product Details