Gene Transcripts

Gene Symbol
Nup54
Entrez Gene ID
53372
Gene Name
nucleoporin 54
Gene Aliases
Chromosome Location
Chr. 14: 17123531 - 17141832 on Build Rnor_6.0
UniGene ID
Rn.11160
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017361.1 NP_059057.1 4, 5 598
GenBank mRNA BC078858.1 - 4, 5 628
U63840.1 - 4, 5 598

View Full Product Details