Gene Transcripts

Gene Symbol
Tcam1
Entrez Gene ID
59305
Gene Name
testicular cell adhesion molecule 1
Gene Aliases
Tcam1p
Chromosome Location
Chr. 10: 94465920 - 94478591 on Build Rnor_6.0
UniGene ID
Rn.125075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021673.1 NP_067705.1 2 310
XM_006247642.2 XP_006247704.1 3 1064
GenBank mRNA BC166495.1 - 3 372

View Full Product Details