Gene Transcripts

Gene Symbol
Pmpcb
Entrez Gene ID
64198
Gene Name
peptidase (mitochondrial processing) beta
Gene Aliases
Mppb
Chromosome Location
Chr. 4: 9908838 - 9921615 on Build Rnor_6.0
UniGene ID
Rn.841
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022395.2 NP_071790.1 5 661
GenBank mRNA BC078826.1 - 5 656
D13907.1 - 5 639
L12965.1 - 5 662

View Full Product Details