Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022399.2 | NP_071794.1 | 2 | 181 | |
GenBank mRNA | BC062395.1 | - | 2 | 158 | |
BU671467.1 | - | 2 | 181 | ||
D78308.1 | - | 2 | 133 | ||
FQ210587.1 | - | 186 | |||
FQ220096.1 | - | 2 | 187 | ||
X53363.1 | - | 2 | 153 | ||
X79327.1 | - | 2 | 131 |