Gene Transcripts

Gene Symbol
SPTBN2
Entrez Gene ID
6712
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
GTRAP41, SCA5, SCAR14
Chromosome Location
Chr. 11: 66682496 - 66729361 on Build GRCh38
UniGene ID
Hs.26915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006946.2 NP_008877.1 7 767
XM_005274192.4 XP_005274249.1 8 1429
XM_005274193.3 XP_005274250.1 7 1203
XM_006718669.3 XP_006718732.1 6 3116
XM_006718671.3 XP_006718734.1 8 829
XM_011545216.2 XP_011543518.1 6 3126
XM_011545217.2 XP_011543519.1 6 3126
XM_017018174.1 XP_016873663.1 8 1521
XM_017018175.1 XP_016873664.1 8 1150
XM_017018176.1 XP_016873665.1 8 870
XM_017018177.1 XP_016873666.1 7 1059
XM_017018178.1 XP_016873667.1 7 742
GenBank mRNA AB008567.1 - 7 767
BC139788.1 - 7 738

View Full Product Details