Gene Transcripts

Gene Symbol
Cul5
Entrez Gene ID
64624
Gene Name
cullin 5
Gene Aliases
Chromosome Location
Chr. 8: 58199992 - 58254195 on Build Rnor_6.0
UniGene ID
Rn.163001
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022683.1 NP_073174.1 9 1105
XM_006243090.2 XP_006243152.1 8 1818
GenBank mRNA AF135115.1 - 9 1105

View Full Product Details