Gene Transcripts

Gene Symbol
Nolc1
Entrez Gene ID
64896
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
Nopp140
Chromosome Location
Chr. 1: 265829057 - 265839819 on Build Rnor_6.0
UniGene ID
Rn.9517
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022869.1 NP_074060.1 2 210
XM_006231560.2 XP_006231622.1 2 261
GenBank mRNA BC126088.1 - 2 175
M94287.1 - 2 210

View Full Product Details