Gene Transcripts

Gene Symbol
Rassf9
Entrez Gene ID
65053
Gene Name
Ras association (RalGDS/AF-6) domain family (N-terminal) member 9
Gene Aliases
P-cip1, Pamci
Chromosome Location
Chr. 7: 44146345 - 44178179 on Build Rnor_6.0
UniGene ID
Rn.30013
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022959.2 NP_075248.1 2 954
GenBank mRNA AF056208.1 - 2 954

View Full Product Details