Gene Transcripts

Gene Symbol
Dab2
Entrez Gene ID
79128
Gene Name
disabled 2, mitogen-responsive phosphoprotein
Gene Aliases
Doc-2, Doc2
Chromosome Location
Chr. 2: 55747353 - 55768848 on Build Rnor_6.0
UniGene ID
Rn.161784
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024159.1 NP_077073.1 9 709
GenBank mRNA BC097314.1 - 9 744
U95177.1 - 9 709

View Full Product Details