Gene Transcripts

Gene Symbol
SSX5
Entrez Gene ID
6758
Gene Name
SSX family member 5
Gene Aliases
Chromosome Location
Chr. X: 48177914 - 48196798 on Build GRCh38
UniGene ID
Hs.166198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021015.3 NP_066295.3 9 1045
NM_175723.1 NP_783729.1 8 922
GenBank mRNA BC016640.2 - 8 1011

View Full Product Details