Gene Transcripts

Gene Symbol
Pggt1b
Entrez Gene ID
81746
Gene Name
protein geranylgeranyltransferase type I, beta subunit
Gene Aliases
Chromosome Location
Chr. 18: 40173236 - 40218225 on Build Rnor_6.0
UniGene ID
Rn.10031
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031082.1 NP_112344.1 4 429
XM_006254691.2 XP_006254753.1 4 1669
XM_008772177.1 XP_008770399.1 5 431
GenBank mRNA L24116.1 - 4 429

View Full Product Details