Gene Transcripts
- Gene Symbol
- Nr2f1
- Entrez Gene ID
- 81808
- Gene Name
- nuclear receptor subfamily 2, group F, member 1
- Gene Aliases
- COUP-TFI
- Chromosome Location
- Chr. 2: 5569954 - 5579894 on Build Rnor_6.0
- UniGene ID
- Rn.89710
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031130.1 |
NP_112392.1
|
1 |
816 |
|
GenBank mRNA |
U10995.1 |
- |
1 |
816 |
|
View Full Product Details