Gene Transcripts

Gene Symbol
Rxrg
Entrez Gene ID
83574
Gene Name
retinoid X receptor gamma
Gene Aliases
Chromosome Location
Chr. 13: 85818473 - 85868555 on Build Rnor_6.0
UniGene ID
Rn.40816
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031765.1 NP_113953.1 3 597
XM_006250212.2 XP_006250274.1 2 741
GenBank mRNA AJ223083.1 - 2 132
BC091363.1 - 3 597 7319081

View Full Product Details