Gene Transcripts

Gene Symbol
Nefl
Entrez Gene ID
83613
Gene Name
neurofilament, light polypeptide
Gene Aliases
NF-L, Nfl
Chromosome Location
Chr. 15: 44799378 - 44803251 on Build Rnor_6.0
UniGene ID
Rn.18568
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031783.1 NP_113971.1 1 895
GenBank mRNA AF031880.1 - 1 895
M25638.1 - 1 233

View Full Product Details