Gene Transcripts

Gene Symbol
Scd2
Entrez Gene ID
83792
Gene Name
stearoyl-Coenzyme A desaturase 2
Gene Aliases
Scd
Chromosome Location
Chr. 1: 264059374 - 264072433 on Build Rnor_6.0
UniGene ID
Rn.83595
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031841.1 NP_114029.1 2 492
GenBank mRNA AB032243.1 - 2 492
BC061737.1 - 2 434 5599134

View Full Product Details