Gene Transcripts

Gene Symbol
Cdh8
Entrez Gene ID
84408
Gene Name
cadherin 8
Gene Aliases
Chromosome Location
Chr. 19: 6031935 - 6423809 on Build Rnor_6.0
UniGene ID
Rn.214220
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053393.2 NP_445845.2 12 2724
XM_008772244.1 XP_008770466.1 11 2071
GenBank mRNA AB010436.1 - 12 2724
AF177679.1 - 1 164

View Full Product Details