Gene Transcripts

Gene Symbol
Folh1
Entrez Gene ID
85309
Gene Name
folate hydrolase 1
Gene Aliases
Naalad
Chromosome Location
Chr. 1: 150323768 - 150395415 on Build Rnor_6.0
UniGene ID
Rn.89278
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057185.2 NP_476533.1 9 1112
GenBank mRNA AF039707.1 - 3 198
AF040256.1 - 9 1118
AF513486.2 - 9 1112
U75973.1 - 9 1069

View Full Product Details