Gene Transcripts

Gene Symbol
Prkcq
Entrez Gene ID
85420
Gene Name
protein kinase C, theta
Gene Aliases
Pkcq
Chromosome Location
Chr. 17: 70971915 - 71105286 on Build Rnor_6.0
UniGene ID
Rn.8227
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276721.1 NP_001263650.1 7 770

View Full Product Details