Gene Transcripts
- Gene Symbol
- Clic5
- Entrez Gene ID
- 94272
- Gene Name
- chloride intracellular channel 5
- Gene Aliases
- Chromosome Location
- Chr. 9: 19121676 - 19372673 on Build Rnor_6.0
- UniGene ID
- Rn.1838
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053603.2 |
NP_446055.1
|
2, 3 |
425 |
|
GenBank mRNA |
AF323174.1 |
- |
2, 3 |
433 |
|
View Full Product Details