Gene Transcripts

Gene Symbol
Csf2
Entrez Gene ID
116630
Gene Name
colony stimulating factor 2 (granulocyte-macrophage)
Gene Aliases
Gm-csf, Gmcsf
Chromosome Location
Chr. 10: 39602089 - 39604070 on Build Rnor_6.0
UniGene ID
Rn.44285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053852.1 NP_446304.1 4 372
GenBank mRNA U00620.1 - 4 321

View Full Product Details