Gene Transcripts
- Gene Symbol
- Csf2
- Entrez Gene ID
- 116630
- Gene Name
- colony stimulating factor 2 (granulocyte-macrophage)
- Gene Aliases
- Gm-csf, Gmcsf
- Chromosome Location
- Chr. 10: 39602089 - 39604070 on Build Rnor_6.0
- UniGene ID
- Rn.44285
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053852.1 |
NP_446304.1
|
1, 2 |
161 |
|
GenBank mRNA |
U00620.1 |
- |
1, 2 |
110 |
|
View Full Product Details