Gene Transcripts

Gene Symbol
TBXA2R
Entrez Gene ID
6915
Gene Name
thromboxane A2 receptor
Gene Aliases
BDPLT13, TXA2-R
Chromosome Location
Chr. 19: 3594506 - 3608749 on Build GRCh38
UniGene ID
Hs.442530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001060.5 NP_001051.1 3 2267
NM_201636.2 NP_963998.2 4 1607
GenBank mRNA BC121148.1 - 2 1526
BC121149.2 - 730
U11271.1 - 4 1432

View Full Product Details