Gene Transcripts

Gene Symbol
Ctrl
Entrez Gene ID
117184
Gene Name
chymotrypsin-like
Gene Aliases
Chromosome Location
Chr. 19: 37905864 - 37907714 on Build Rnor_6.0
UniGene ID
Rn.63352
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054009.1 NP_446461.1 2 90
GenBank mRNA AB020757.1 - 2 90
FQ234836.1 - 2 102

View Full Product Details