Gene Transcripts

Gene Symbol
Csrp3
Entrez Gene ID
117505
Gene Name
cysteine and glycine-rich protein 3 (cardiac LIM protein)
Gene Aliases
MLP
Chromosome Location
Chr. 1: 104147205 - 104166389 on Build Rnor_6.0
UniGene ID
Rn.11345
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057144.1 NP_476485.1 3 371
XM_006229237.2 XP_006229299.1 4 475
GenBank mRNA X81193.1 - 3 371

View Full Product Details