Gene Transcripts

Gene Symbol
Siah2
Entrez Gene ID
140593
Gene Name
siah E3 ubiquitin protein ligase 2
Gene Aliases
Chromosome Location
Chr. 2: 148874151 - 148891900 on Build Rnor_6.0
UniGene ID
Rn.29192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134457.1 NP_604452.1 1 489
GenBank mRNA AB067815.1 - 1 489
AF389477.1 - 1 43

View Full Product Details