Gene Transcripts

Gene Symbol
Rcan2
Entrez Gene ID
140666
Gene Name
regulator of calcineurin 2
Gene Aliases
Dcip2, Dscr1l1, Zaki-4
Chromosome Location
Chr. 9: 19518568 - 19749145 on Build Rnor_6.0
UniGene ID
Rn.24428
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175578.3 NP_783168.1 2, 3 454
GenBank mRNA AF459023.1 - 2, 3 296
BC086974.1 - 2, 3 454 7193921

View Full Product Details