Gene Transcripts

Gene Symbol
Mgea5
Entrez Gene ID
154968
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
Gene Aliases
Ncoat, O-GLCNACASE, OGA
Chromosome Location
Chr. 1: 265508619 - 265541300 on Build Rnor_6.0
UniGene ID
Rn.162539
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131904.1 NP_571979.1 7 933
GenBank mRNA AY039679.1 - 7 933

View Full Product Details