Gene Transcripts

Gene Symbol
Hadha
Entrez Gene ID
170670
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
RGD1560655
Chromosome Location
Chr. 6: 27589840 - 27628921 on Build Rnor_6.0
UniGene ID
Rn.3340
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130826.2 NP_570839.2 8 728
GenBank mRNA BC091697.1 - 8 728
D16478.1 - 8 726

View Full Product Details