Gene Transcripts

Gene Symbol
Slc40a1
Entrez Gene ID
170840
Gene Name
solute carrier family 40 (iron-regulated transporter), member 1
Gene Aliases
Fpn1, Slc11a3, Slc39a1
Chromosome Location
Chr. 9: 52894365 - 52912293 on Build Rnor_6.0
UniGene ID
Rn.15324
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133315.2 NP_579849.2 7 1191
GenBank mRNA AF394785.3 - 7 1136
CV104407.1 - 2 263
FQ226522.1 - 6 851
U76714.1 - 7 1206

Gene Symbol
LOC100911874
Entrez Gene ID
100911874
Gene Name
solute carrier family 40 member 1-like
Gene Aliases
Chromosome Location
Chr. 9: 52819386 - 52831441 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750695.3 XP_003750743.1 1614

View Full Product Details