Gene Transcripts

Gene Symbol
Tnrc6b
Entrez Gene ID
192178
Gene Name
trinucleotide repeat containing 6B
Gene Aliases
Cbl27
Chromosome Location
Chr. 7: 121930638 - 122147934 on Build Rnor_6.0
UniGene ID
Rn.17120
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138845.2 NP_620200.2 8 1052
XM_006242061.2 XP_006242123.1 4 1161
XM_006242063.2 XP_006242125.1 4 1160
XM_006242064.2 XP_006242126.1 4 1160

View Full Product Details