Gene Transcripts

Gene Symbol
Otos
Entrez Gene ID
246044
Gene Name
otospiralin
Gene Aliases
Chromosome Location
Chr. 9: 99820202 - 99824115 on Build Rnor_6.0
UniGene ID
Rn.56698
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139188.3 NP_631927.1 4 209
XM_008767358.1 XP_008765580.1 5 817
XM_008767359.1 XP_008765581.1 4 575
GenBank mRNA AY062254.1 - 4 237

View Full Product Details