Gene Transcripts

Gene Symbol
Ctsq
Entrez Gene ID
246147
Gene Name
cathepsin Q
Gene Aliases
CatQ
Chromosome Location
Chr. 17: 3991330 - 3996889 on Build Rnor_6.0
UniGene ID
Rn.34875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139262.1 NP_640355.1 7 838
GenBank mRNA AF187323.1 - 7 838

View Full Product Details