Gene Transcripts
- Gene Symbol
- Prpmp5
- Entrez Gene ID
- 257651
- Gene Name
- proline-rich protein MP5
- Gene Aliases
- Prb1, Prpg2
- Chromosome Location
- Chr. 4: 166013226 - 166016335 on Build Rnor_6.0
- UniGene ID
- Rn.29950
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172065.2 |
NP_742062.1
|
1, 3 |
94 |
|
GenBank mRNA |
L17318.1 |
- |
1, 3 |
95 |
|
View Full Product Details