Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004617.3 | NP_004608.1 | 1 | 446 | |
GenBank mRNA | AK055577.1 | - | 1 | 1053 | |
AK315464.1 | - | 1 | 333 | ||
BC001386.2 | - | 1 | 316 | 3051146 | |
BP294159.1 | - | 1 | 446 | ||
BT007430.1 | - | 1 | 149 | ||
U31449.1 | - | 1 | 314 |