Gene Transcripts

Gene Symbol
Tbc1d24
Entrez Gene ID
287110
Gene Name
TBC1 domain family, member 24
Gene Aliases
RGD1306143
Chromosome Location
Chr. 10: 13551100 - 13576739 on Build Rnor_6.0
UniGene ID
Rn.224964
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105769.1 NP_001099239.1 4, 5 1401
XM_006245926.1 XP_006245988.1 3, 4 1160

View Full Product Details