Gene Transcripts

Gene Symbol
Metrn
Entrez Gene ID
287151
Gene Name
meteorin, glial cell differentiation regulator
Gene Aliases
Hyrac, RGD1306222
Chromosome Location
Chr. 10: 15164439 - 15166457 on Build Rnor_6.0
UniGene ID
Rn.50391
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009962.1 NP_001009962.1 2 422
GenBank mRNA AY800384.1 - 2 422

View Full Product Details